Tropaeolin O
About this item
Tropaeolin O, also known as Chrysoine resorcinol or Yellow T, is a synthetic azo dye used as a pH indicator.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
- CAS Number:547-57-9
- Formula:C₁₂H₉N₂O₅SNa
- MDL Number:MFCD00007499
- Molecular Weight:316.26 g/mol
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
Tropaeolin O, also known as Chrysoine resorcinol or Yellow T, is a synthetic azo dye used as a pH indicator.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
- CAS Number:547-57-9
- Formula:C₁₂H₉N₂O₅SNa
- MDL Number:MFCD00007499
- Molecular Weight:316.26 g/mol
- Storage Temperature:Ambient
Documents
TR145_spec.pdf
Product specification
Specifications for Product Family
Specification Test Results
- Absorption Max:387 NM
- Visual Transition Intervals:pH 11.0 (Yellow)
Product Family Options
Product Information
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



