Order Entry
Puerto Rico
ContactUsLinkComponent
L(+)-Lysine monohydrochloride ≥98.5%, granular USP
L(+)-Lysine monohydrochloride ≥98.5%, granular USP
Catalog # 95034-130
Small Business Enterprise
CAS Number:  
L(+)-Lysine monohydrochloride ≥98.5%, granular USP
Catalog # 95034-130
Supplier Number:  L1142-100GM
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Poly bottle
  • Size:
    100 g
  • Cat. No.:
    95034-130
  • Supplier no.:
    L1142-100GM

Specifications

Specification Test Results

  • Assay (Dried Basis):
    98.5 - 101.5%
  • Specific Rotation [A]D:
    +20.4 to +21.4°
  • Chloride Content:
    19.0 - 19.6%
  • Related Compounds:
    To Pass Test
  • Loss On Drying:
    max. 0.4%

Specification Test Results

About this item

 :
 :182.65 g/mol
Storage Temperature:Ambient
CAS Number:657-27-2
Formula:H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
MDL Number:MFCD00064564