L-Alanyl-L-glutamine is dipeptide formed from Alanine and Glutamine and is a stable and soluble delivery form of glutamine, an amino acid. In cosmetics and personal care products, it functions primarily as a humectant and an emollient skin conditioning agent. It can be found in skin care products. It appears as a white to off-white powder that is soluble in water.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- CAS No.:39537-23-0
- Storage Conditions:Ambient
- MDL No.:MFCD00133046
- Protein Synonyms:H-Ala-Gln-OH
- Protein/Peptide Name:L-Alanyl-L-glutamine
- Purity:>98.0% (HPLC,T)
- Molecular Weight:217.23 g/mol
- Molecular Formula:C₈H₁₅N₃O₄