DL-Mevalonolactone
About this item
DL-Mevalonic Acid Lactone, an organic compound in biochemistry, is the primary precursor of isopentenyl pyrophosphate.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
- CAS Number:674-26-0
- Formula:C₆H₁₀O₃
- MDL Number:MFCD00006648
- Molecular Weight:130.14 g/mol
- Storage Temperature:Freezer
2 Options Available Below
Product Details & Documents
DL-Mevalonic Acid Lactone, an organic compound in biochemistry, is the primary precursor of isopentenyl pyrophosphate.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
- CAS Number:674-26-0
- Formula:C₆H₁₀O₃
- MDL Number:MFCD00006648
- Molecular Weight:130.14 g/mol
- Storage Temperature:Freezer
Product Family Options
Product Information
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



