About this item
Aspartame, Powder, NF is used in pharmaceutical products, often as a sugar replacement in chewable tablets and sugar-free liquids. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
- Transmittance at 430 nm: To pass test
- Specific rotation [A]D: +14.5 to +16.5
- Loss On drying: Max. 4.5%
- Residue on ignition: Max. 0.2%
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- CAS No.:22839-47-0
- Storage Conditions:Ambient
- Form:Powder
- MDL No.:MFCD00002724
- Protein Synonyms:H-Asp-Phe-OMe|(3S)-3-amino-4-[[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]amino]-4-oxobutanoic acid|L-AspartyL-L-phenylalanine methyl ester|N-L-α-Aspartyl-L-phenylalanine 1-Methyl Ester
- Protein/Peptide Name:Aspartame
- Purity:98.0% (Dried basis)
- Molecular Weight:294.30 g/mol
- Grade:NF
- Molecular Formula:C₁₄H₁₈N₂O₅