Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
2',7'-Dichlorofluorescein ACS

2',7'-Dichlorofluorescein ACS

2',7'-Dichlorofluorescein ACS
Certificates

4 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

2',7'-Dichlorofluorescein, Reagent, ACS, an organic dye of the fluorescein family, is used in the cellular antioxidant activity assay.

As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Synonyms:  2',7'-Dichloro-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one
  • CAS Number:76-54-0
  • Formula:C₂₀H₁₀Cl₂O₅
  • MDL Number:MFCD00005047
  • Molecular Weight:401.20 g/mol
  • Storage Temperature:Ambient

Documents

Certificates
Search by Lot Number
Specifications for Product Family

Specification Test Results

  • Clarity of Alcohol Solution:
    To Pass Test
  • Suitability As Adsorption Indicator:
    To Pass Test
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass, amber
    1 g
    Log in for availability
    Log in for availability
    Each (1g)
    Each (1g) - $82.41
    $82.41
    Each (1g)
       $82.41

    Supplier Number: DI116-1GM
  • Glass, amber
    5 g
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $241.57
    $241.57
    Each (5g)
       $241.57

    Supplier Number: DI116-5GM
  • Glass, amber
    25 g
    Log in for availability
    Log in for availability
    Each (25g)
    Each (25g) - $716.24
    $716.24
    Each (25g)
       $716.24

    Supplier Number: DI116-25GM
  • Glass, amber
    500 g
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $3,509.12
    $3,509.12
    Each (500g)
       $3,509.12

    Supplier Number: DI116-500GMB

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.