Micropipette Puller, P-1000, Sutter Instrument
Catalog # 102966-584
Supplier: Sutter Instruments Company
Specifications
- Description:Next Generation Micropipette Puller, P-1000
- Dimensions:21×14×12"
- Cat. No.:102966-584
- Electrical:115/230 V, 50/60 Hz
- Weight:18.59
- Supplier no.:P1000
Specifications
About this item
The P-1000 Next Generation Micropipette Pullers features a color touchscreen display that provides an intuitive and full-featured interface.
- Safe heat mode to protect and extend filament life
- Jaw temperature sensor helps define ideal pulling conditions
- Self-contained air supply with filtration system and humidity control chamber
- Memory storage for up to 100 programs
- Two cooling modes - time and delay
Diagnostic testing of all puller components with built-in error detection of air pressure loss or filament burnout, easy access to ramp test, measurement of jaw temperature, and access to previous pull results with the heat on times for each cycle of the program. The topics are pre-loaded to assist with on-site troubleshooting and the built-in glossary includes text, pictures, and diagrams explaining the terms used in micropipette fabrication.