
Acetic acid 2.0 M CH3COOH (2.0N) volumetric solution, Fluka™
Catalog # BJA8976-100ML
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:100 ml
- Cat. No.:BJA8976-100ML
- Supplier no.:A8976-100ML
Specifications
Specification Test Results
- Assay:1.95 - 2.05 N
Specification Test Results
About this item
:
:60.05 g/mol
Storage Temperature:Ambient
UN:2790
ADR:8,III
:GHS05
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152