ContactUsLinkComponent
VWR® Wide Mouth Bottles, HDPE
VWR® Wide Mouth Bottles, HDPE
Catalog # 75800-482
Supplier:  VWR International
VWR® Wide Mouth Bottles, HDPE
Catalog # 75800-482
Supplier:  VWR International
Specifications
  • Capacity:
    500 ml (16.9 oz.)
  • Material:
    HDPE
  • Cat. No.:
    75800-482
Specifications
About this item
These bottles offer reliable durability and excellent chemical resistance.
  • Bottles are used for packing, shipping, and storage purposes
  • Fill line marked
Delivery: Supplied with leak-proof polypropylene screw cap.