ContactUsLinkComponent
Prolystica™ 2X Enzymatic Presoak and Cleaner, STERIS®
Prolystica™ 2X Enzymatic Presoak and Cleaner, STERIS®
Catalog # 89176-724
Supplier:  Steris
Prolystica™ 2X Enzymatic Presoak and Cleaner, STERIS®
Catalog # 89176-724
Supplier:  Steris
Supplier Number:  1C3308
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Prolystica™ 2X Enzymatic Presoak and Cleaner
  • Size:
    3.78 L (1 gal.)
  • Cat. No.:
    89176-724
  • Supplier no.:
    1C3308
Specifications
About this item
Prolystica™ Enzymatic Presoak and Cleaner is a dual enzyme formula which provides exceptional proteolytic Performance within a broad range of water quality and types.
  • Enhanced cleaning of fatty orthopedic and protein soils in multiple applications
  • Built-in protection against instrument damage
  • Pleasant new fragrance and biodegradable
Fast action provides improved productivity and quicker instrument turn-around for facility. Low foaming characteristics allows for maximum washer efficiency and rinse ability.