Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Heavy Duty Orbital Shaker, Eberbach

Heavy Duty Orbital Shaker, Eberbach

115 V, 50/60 Hz
Catalog # 75805-250
Supplier:   Eberbach
Supplier #:  E6155.00
Heavy Duty Orbital Shaker, Eberbach
Catalog # 75805-250
Supplier:  Eberbach
Heavy Duty Orbital Shaker, Eberbach
Heavy Duty Orbital Shaker, Eberbach

115 V, 50/60 Hz

Price
$53,591.02
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

One of the highest capacity orbital shakers on the market. With ability to shake up to 500 lbs of sample. Eberbach’s proprietary closed loop controller ensures the most accurate mixer speeds at all rpms; if accuracy and repeatability is what you require Eberbach has the best in class! Also, featuring a built in pallet jack base for easy transportation and placement.

  • 500 lbs. weight capacity
  • 30 to 300 rpm
  • Digital controller
  • 5 year warranty

Eberbach’s large capacity Variable-Speed Orbital Shakers are designed for rotary swirling action and is used for bioprocessing of plant cell cultures, large batch mammalian cell culturing, aeration of fermentations and various other chemical mixing procedures.

The orbital motion of the E6155 ensures efficient liquid mixing. The E6155 allows high oxygen transfer rates with low shear forces not seen with other bio-reactors which use propellers or impellers. The E6155 reduces stress on cells thanks to the swirling action of the shaker. This motion is particularly suitable for cell cultivation and fermentation. The E6155 uses Eberbach unique balanced bearing plates for a long lasting smooth operation, repeatable motion with accurate control of the orbital speed and time duration.

When used with optional biochemical sensors, the E6155 is ideal for scale-up fermentation, cultivation and bio processing applications. The base is designed to be moved with a forklift or pallet jack. The E6155 uses Eberbach’s own controller which has a digital tachometer along with digital countdown timer and optional audible end of cycle alarm. The digital countdown timer can be selected for timed operation or turned off for constant operation.  The eccentric drive is counterbalanced to assure smooth acceleration and minimal bearing wear for increased life of the machine.  Speed is continuously variable between from 30 - 300 rpm or with the Eberbach controller you can program your specific speed. The E6155 comes factory set with a 2” (5 cm) orbit, if your application requires a different orbit size we can offer alternative sizes.

Accessories information: The E6155 can be supplied with multiple optional features; remote control box mounted to a pendant with castors or as a wall mount box, Sanitary models (stainless steel with IP67 rated motor), hazardous location models (explosion-proof), custom top platforms and more. Contact your Avantor representative for more information.

Ordering information: Box Carrier is not included (Box Carrier E6155.A.UB pictured). Accessories not included.

Packaging: Product ships LTL in a crate.

Specifications
  • Electrical:
    115 V, 50/60 Hz
  • Cat. No.:
    75805-250
  • Speed:
    30 - 300 rpm
  • Orbit:
    2"
  • Shipping Weight:
    2400 lbs.
  • Max. Load Capacity:
    50 - 500 lbs.
  • Dimensions:
    43 x 43 x 20"
  • Shipping Dimensions:
    49 x 49 x 45"