Microscale Slide, Watson Bio Lab
Microscale slide
Catalog # 75982-988
Supplier: Watson Bio Lab
Supplier #: 177-401C
Jump to:
Product Details & Documents
This polystyrene slide has smooth scale lines.
- Accurate Linearity
- Inexpensive
- Polystyrene
- Smooth Scale Lines
Users can apply the sample directly on the Microscale when observing with a microscope
Please replace with a new slide after using several times and if it is hard to see due to dirt and scratch.
The economic slide is less expensive than a traditional glass slide.
Documents
micro_scale
Videos
Specifications
- Description:Microscale slide