Hyper Microtubes, Watson Bio Lab
About this item
These hyper microtubes feature a writing space, a locking mechanism, and a friendly hinge for fingers.
- Autoclavable Polypropylene
- Writing Space
- Locking Mechanism
- Hinge for Fingers
- Scale Graduations
The flat top allows for marking on the microtubes' top, and the surface of the tube itself offers more writing space.
The round-bottom microtubes feature autoclavable polypropylene construction. A scale marks each 0.5 ml on the tubes. The lock part prevents the thumb from touching the inner cap.
Accessories information: Pestles, storage racks, and cap locks are available separately.
3 Options Available Below
Product Details & Documents
These hyper microtubes feature a writing space, a locking mechanism, and a friendly hinge for fingers.
- Autoclavable Polypropylene
- Writing Space
- Locking Mechanism
- Hinge for Fingers
- Scale Graduations
The flat top allows for marking on the microtubes' top, and the surface of the tube itself offers more writing space.
The round-bottom microtubes feature autoclavable polypropylene construction. A scale marks each 0.5 ml on the tubes. The lock part prevents the thumb from touching the inner cap.
Accessories information: Pestles, storage racks, and cap locks are available separately.
Product Family Options
Product Information