Orders LinkContactUsLinkComponent
TransforMax™EC100™ Electrocompetent and Chemically Competent E. coli, Biosearch Technologies
Supplier:   Lucigen
TransforMax™EC100™ Electrocompetent and Chemically Competent <i>E. coli</i>, Biosearch Technologies
Supplier:  Lucigen
Certificates
TransforMax™EC100™ Electrocompetent and Chemically Competent <i>E. coli</i>, Biosearch Technologies
TransforMax™EC100™ Electrocompetent and Chemically Competent E. coli, Biosearch Technologies
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Clone large plasmids with high efficiency.
  • Clone large inserts and transform large plasmids - up to at least 145 kb plasmid DNA
  • Select convenience with chemically competent format: >5 x 108 cfu/µg pUC19 DNA
  • Choose electrocompetent format for higher efficiencies: >1 x 1010 cfu/µg pUC19 DNA
  • Ideal strain for general cloning, subcloning and plasmid preparation due to endA and recA mutation
The highly versatile TransforMax™ EC100™ E
EC100 competent cells are recombination minus (recA) and endonuclease minus (endA), for high yield, high quality plasmid DNA preparations. They are also blue/white screening capable (lacZΔM15) and restriction minus [mcrA, Δ(mrr-hsdRMS-mcrBC)] for cloning methylated DNA.
Genotype: F- mcrA Δ(mrr-hsdRMS-mcrBC) Φ80dlacZΔM15 ΔlacX74 recA1 endA1 araD139 Δ(ara, leu)7697 galU galK λ- rpsL (StrR) nupG.
coli competent cells are ideal for most cloning applications. The cells provide very high transformation efficiency when tested against a wide range of supercoiled DNAs as well as DNA directly from a ligation reaction, including BAC clones.
 : TransforMax™ EC100™ Chemically Competent E. coli kit contains ten tubes, each containing 50 µL of cells (enough cells for 10 transformations) and pUC19 control DNA.
TransforMax™ EC100™ Electrocompetent E. coli kit contains ten tubes, each containing 100 µL of cells (enough cells for 20 transformations) and pUC19 control DNA.
 : For research use only. Not for human or diagnostic use.
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 75927-880
    TransforMax™ EC100™ Chemically Competent E. coli
    TransforMax™ EC100™ Chemically Competent E. coli
    10 x 50 µL
    Log in for availability
    Log in for availability
    Each
    Each - $341.22
    $341.22
    Each
       $341.22
    Supplier Number: CC02810
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 75927-926
    TransforMax™ EC100™ Electrocompetent E. coli
    TransforMax™ EC100™ Electrocompetent E. coli
    10 x 100 µL
    Log in for availability
    Log in for availability
    Each
    Each - $718.28
    $718.28
    Each
       $718.28
    Supplier Number: EC10010
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.