To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Peroxigen ST Sterile, 6% Hydrogen Peroxide Solution
- Size:16 oz.
- Environmentally Preferable:Sustainable Packaging,Waste Reducing
- Cat. No.:75853-810
- Packaging:Trigger spray bottle
- Supplier no.:9716
Specifications
About this item
Peroxigen ST is a sterile 6% (v/v) Hydrogen Peroxide solution made with water-for-injection (WFI).
Peroxigen ST is filtered to 0.2 microns
- Bottled and double-bagged within a ISO CLASS 5 cleanroom
- Gamma-irradiated to a sterility assurance level (SAL) of 10⁻⁶
This ready-to-use solution can be used to clean surfaces and equipment found in cleanroom areas within pharmaceutical, biotechnology, and medical device facilities, or anywhere the use of a sterile, hydrogen peroxide solution is required.
Each lot is tested for sterility via USP 14-day sterility test. A QR code can be found on each case of Peroxigen ST which is linked to a copy of the lot specific document, which details QC, irradiation, and sterility for that particular lot.