Order Entry
Puerto Rico
ContactUsLinkComponent
MMLV Reverse Transcriptase 1st-Strand cDNA Synthesis Kit, Biosearch Technologies
MMLV Reverse Transcriptase 1st-Strand cDNA Synthesis Kit, Biosearch Technologies
Catalog # 75928-118
Supplier:  Lucigen
CAS Number:  
undefined
MMLV Reverse Transcriptase 1st-Strand cDNA Synthesis Kit, Biosearch Technologies
Catalog # 75928-118
Supplier:  Lucigen
Supplier Number:  MM070150
CAS Number:  

Specifications

  • Description:
    MMLV Reverse Transcriptase 1st-Strand cDNA Synthesis Kit
  • Cat. No.:
    75928-118
  • No. of reactions:
    50
  • Supplier no.:
    MM070150

Specifications

About this item

An optimized, complete system for the production of full-length cDNA from low amounts of total RNA.

  • Robust: Produce full-length cDNA >15 kb long
  • Sensitive: Synthesize cDNA from picogram amounts of starting total RNA
  • Flexible: Includes both random nonamer and oligo-dT primers
  • Complete: Includes all components necessary as well as the RiboGuard™ RNase Inhibitor to protect your RNA before reverse transcription and increase cDNA yield

This kit is used for first-strand cDNA synthesis which can be followed by PCR or qPCR

The MMLV Reverse Transcriptase 1st-Strand cDNA Synthesis Kit produces full-length first-strand cDNA from total cellular RNA preparations or purified poly(A) RNA.

This complete kit includes the same MMLV High Performance Reverse Transcriptase that is available as a stand alone reverse transcriptase and maintains all the same high performance characteristics as that enzyme.

cDNA synthesized with this kit is useful for a variety of downstream applications including (i) RT-PCR validation of gene expression data obtained from microarray or next gen sequencing experiments, and (ii) RT-PCR validation and quantification of gene silencing by RNA interference.

Delivery: Each kit contains MMLV Reverse Transcriptase, 10X Reaction Buffer, RiboGuard™ RNase Inhibitor, dNTP PreMix, DTT, Oligo(dT)21 Primer, Random 9-mer Primers and RNase-free Water.
Caution: For research use only. Not for human or diagnostic use.