MMLV High Performance Reverse Transcriptase, cDNA Synthesis, Biosearch Technologies
MMLV High Performance Reverse Transcriptase, 25000 Units
Catalog # 75927-968
Supplier: Lucigen
Supplier #: RT80125K
MMLV High Performance Reverse Transcriptase, cDNA Synthesis, Biosearch Technologies
Catalog # 75927-968
Supplier: Lucigen
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
An optimized reverse transcriptase and buffer system for the production of full-length cDNA.
- Robust: Produce full-length cDNA >15 kb long
- Sensitive: Produce cDNA from picogram amounts of starting RNA
- Flexible: Optimize the RT reaction to your specific needs by using the primers, dNTPs and RNase inhibitor of your choice (not included)
The MMLV High Performance Reverse Transcriptase Enzyme demonstrates significantly greater reverse transcriptase activity than other commercially available MMLV RT enzymes. Typically, just 100 units of MMLV HP RT are required for full-length cDNA synthesis compared to 200 units of competitive MMLV RT enzymes. MMLV HP RT includes a 10X Reaction Buffer, optimized for synthesis of full-length cDNA from long RNA templates, and DTT.
: Contains MMLV High Performance Reverse Transcriptase, 10X Reaction Buffer and DTT.
: For research use only. Not for human or diagnostic use.
Specifications
Specifications
- Description:MMLV High Performance Reverse Transcriptase
- Size:25000 Units