Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Coating solution
Coating solution concentrate for ELISA
Coating solution concentrate for ELISA
Coating solution concentrate for ELISA
Catalog # 95059-194
Supplier:
KPL
CAS Number:
Expand image
Coating solution concentrate for ELISA
Catalog # 95059-194
Supplier:
KPL
Supplier Number:
5150-0014
CAS Number:
Specifications
Size:
50 ml
Cat. No.:
95059-194
Supplier no.:
50-84-00
Specifications
About this item
Product Details
:
Storage Temperature:
Refrigerator
Product Details