Order Entry
Puerto Rico
ContactUsLinkComponent
 
Pierce™ TCEP Solution, Neutral pH, Bond-Breaker™, Thermo Scientific
  PI77720
 :  
undefined
Pierce™ TCEP Solution, Neutral pH, Bond-Breaker™, Thermo Scientific
  PI77720
 :  77720
 :  

 

  • Description:
    Bond-Breaker® Tris[2-Carboxyethyl]phosphine Neutral Solution (TCEP)
  • Size:
    5 mL
  • Cat. No.:
    PI77720
  • Molecular weight:
    286.65 g/mol
  • Application:
    Water-Soluble Reducing Agent
  • Supplier no.:
    77720

 

 

Bond-Breaker™ TCEP Solution (neutral pH) is a stable, 0.5 M solution of the thiol-free, phosphine-based TCEP compound, useful as a 10X stock for addition to SDS-PAGE sample loading buffers to reduce protein disulphide bonds.

  • Odourless
  • Selective and complete reduction of even the most stable water-soluble alkyl disulfides
  • Reduces protein disulfides at room temperature and pH 5 in less than five minutes
  • Resistant to air oxidation; nonvolatile and nonreactive toward other functional groups found in proteins