Pierce™ DyLight™ Antibody Labeling, NHS Ester Quenchers, Thermo Scientific
PI84512
:
- Description:DyLight™ 425Q NHS ester
- Size:1 mg
- Storage Conditions:Store at 4˚C.
- Cat. No.:PI84512
- Supplier no.:84512
DyLight™ NHS Ester Quenchers are amine-reactive non-fluorescent dyes that use fluorescent resonance energy transfer (FRET) to quench fluorescent signals resulting from ultraviolet, visible and infrared light-excited fluorescent dyes
These dyes can be efficiently paired with many DyLight™ dyes and other common fluorescent dyes including fluorescein, rhodamine, cyanine and Texas Red® dye.