Order Entry
Puerto Rico
ContactUsLinkComponent
tert-Butyl methyl ether ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Flammable
Irritant
tert-Butyl methyl ether ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Catalog # BJ650560-1L
CAS Number:  
undefined
Flammable
Irritant
tert-Butyl methyl ether ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Catalog # BJ650560-1L
Supplier Number:  650560-1L
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 L
  • Cat. No.:
    BJ650560-1L
  • Supplier no.:
    650560-1L

Specifications

Specification Test Results

  • Purity (GC):
    Min. 99.85 %
  • IR spectrum:
    complying
  • Non-volatile matter:
    Max. 0.0001 %
  • Water (Karl Fischer):
    Max. 0.02 %
  • Absorbance at 400 nm:
    Max. 0.005
  • Absorbance at 300 nm:
    Max. 0.005
  • Absorbance at 250 nm:
    Max. 0.10
  • Absorbance at 225 nm:
    Max. 0.50
  • Absorbance at 210 nm:
    Max. 1.0
  • Fluorescence (chinin) at 365 nm:
    Max. 1.0 ppb
  • GC (as heptachlor epoxide):
    Max. 10 ng/l
  • Refractive Index:
    n20/D 1.369 (lit.)
  • Autoignition Temperature:
    705 °F

Specification Test Results

About this item

 :
 :88.15 g/mol
Boiling Pt:55.3 °C (1.013 hPa)
Melting Pt:–109 °C
Density:0.74 g/cm³ (20 °C)
Flash Pt:8.5 %(V)
Storage Temperature:Ambient
EINECS:216-653-1
UN:2398
ADR:3,II
 :GHS02
 :GHS07
CAS Number:1634-04-4
Formula:(CH₃)₃COCH₃
MDL Number:MFCD00008812