

Chloroform ≥99.9% stabilized, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Catalog # BJ650498-1L
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:1 L
- Cat. No.:BJ650498-1L
- Supplier no.:650498-1L
Specifications
Specification Test Results
- Identity (IR):complying
- Assay (GC):Min. 99.90 %
- Non-volatile matter:Max. 0.0001 %
- Water (Karl Fischer):Max. 0.02 %
- GC (as heptachlor epoxide):Max. 10 ng/l
- 2-methyl-2-buten (GC):20 - 200 ppm
- Absorbance at 400 nm:Max. 0.005
- Absorbance at 275 nm:Max. 0.005
- Absorbance at 250 nm:Max. 0.30
- Absorbance at 245 nm:Max. 1.00
- Fluorescence (chinin) at 365 nm:Max. 0.5 ppb
- Contains:amylenes as stabilizer
- Refractive Index:n20/D 1.445 (lit.)
Specification Test Results
About this item
:
:119.38 g/mol
Boiling Pt:61 °C (1.013 hPa)
Melting Pt:–61.3 °C
Density:1.485 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-663-8
UN:1888
ADR:6.1,III
:Stabilised with amylene
:GHS07
:GHS08
CAS Number:67-66-3
Formula:CHCl₃
MDL Number:MFCD00000826