Order Entry
Puerto Rico
ContactUsLinkComponent
 
Dichloromethane ≥99.9% stabilized, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
 
undefined
Dichloromethane ≥99.9% stabilized, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
 

 

  • Appearance (color):
    complying
  • Appearance (form):
    complying
  • Assay (GC):
    Min. 99.9 %
  • IR spectrum:
    complying
  • Non-volatile matter:
    Max. 0.0001 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Absorbance at 400 nm:
    Max. 0.005
  • Absorbance at 300 nm:
    Max. 0.005
  • Absorbance at 250 nm:
    Max. 0.02
  • Absorbance at 240 nm:
    Max. 0.10
  • Absorbance at 235 nm:
    Max. 1.00
  • Fluorescence (chinin) at 365 nm:
    Max. 1 ppb
  • 2-methyl-2-buten (GC):
    50 - 150 ppm
  • Suitability for residue analysis:
    Max. 10 ng/l
  • Vapor Pressure:
    24.45 psi ( 55 °C)