


Cyclohexane ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Catalog # BJ650455-1L
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:1 L
- Cat. No.:BJ650455-1L
- Supplier no.:650455-1L
Specifications
Specification Test Results
- IR spectrum:complying
- Assay (GC):Min. 99.90 %
- Non-volatile matter:Max. 0.0001 %
- Water (Karl Fischer):Max. 0.01 %
- Absorbance at 400 nm:Max. 0.005
- Absorbance at 300 nm:Max. 0.005
- Absorbance at 250 nm:Max. 0.02
- Absorbance at 225 nm:Max. 0.17
- Absorbance at 200 nm:Max. 1.00
- Fluorescence (chinin) at 365 nm:Max. 1 ppb
- GC (as heptachlor epoxide):Max. 10 ng/l
- Explosion Limit:9 %
- Autoignition Temperature:500 °F
Specification Test Results
About this item
:
:84.16 g/mol
Boiling Pt:81 °C (1.013 hPa)
Melting Pt:6.4 °C
Density:0.78 g/cm³ (20 °C)
Flash Pt:8.3 %(V)
Storage Temperature:Ambient
EINECS:203-806-2
UN:1145
ADR:3,II
:GHS02
:GHS08
:GHS07
CAS Number:110-82-7
Formula:C₆H₁₂
MDL Number:MFCD00003814