Order Entry
Puerto Rico
ContactUsLinkComponent
 
 
 
Chloroform ≥99.8% stabilized, CHROMASOLV™ for HPLC, Riedel-de Haën™
  BJ366927-4X4L
 :  
undefined
 
 
Chloroform ≥99.8% stabilized, CHROMASOLV™ for HPLC, Riedel-de Haën™
  BJ366927-4X4L
 :  366927-4X4L
 :  

 

  • Pack type:
    Glass bottle
  • Size:
    4 L
  • Cat. No.:
    BJ366927-4X4L

 

 

  • Identity (IR):
    complying
  • Purity excluding stabilizer (GC):
    min. 99.80 %
  • Ethanol (GC):
    0.5 - 1.0 %
  • Non-volatile matter:
    max. 0.0003 %
  • Water (Karl Fischer):
    max. 0.01 %
  • Free acid and chloride:
    complying
  • Lead (Pb):
    max. 0.05 ppm
  • Free chlorine:
    complying
  • Reaction against H2SO4:
    complying
  • Acetone, aldehydes (neßler test):
    complying
  • Absorbance at 400 nm:
    max. 0.01
  • Absorbance at 290 nm:
    max. 0.01
  • Absorbance at 270 nm:
    max. 0.02
  • Absorbance at 260 nm:
    max. 0.15
  • Absorbance at 255 nm:
    max. 0.15
  • Absorbance at 245 nm:
    max. 1.00
  • APHA:
    max. 10

 

 

 :
 :119.38 g/mol
 :62 °C
 :–64 °C
 :1.49 g/cm³ (20 °C)
 : Does not flash
 :1888
 :6.1,III
 :Stabilised with ethanol 0.5-1.0%
 :GHS07
 :GHS08
 :Ambient
 :67-66-3
 :CHCl₃
 :MFCD00000826