

Ammonia solution 28% (NH₃ basis) in water ≥99.99% (trace metals basis)
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MDL Number:MFCD00066650
:35.05 g/mol
Boiling Pt:32 °C (1.013 hPa)
Melting Pt:–92 °C
Density:0.89 g/cm³ (20 °C)
Flash Pt:30.2 %(V)
Storage Temperature:Ambient
UN:2672
ADR:8,III
:GHS07
:GHS05
Specification Test Results
- Assay (acidimetric):28.0 - 30.0 %
- Metal trace analysis (ICP):Max. 0.01 %