Order Entry
Puerto Rico
ContactUsLinkComponent
Starch 1% in water stabilized indicator, Fluka™
Starch 1% in water stabilized indicator, Fluka™
Catalog # BJ319554-500ML
CAS Number:  
Starch 1% in water stabilized indicator, Fluka™
Catalog # BJ319554-500ML
Supplier Number:  319554-500ML
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    500 ml
  • Cat. No.:
    BJ319554-500ML
  • Supplier no.:
    319554-500ML

Specifications

Specification Test Results

  • Turbidity:
    Max. 50 NTU
  • Test of sensitivity:
    complying

Specification Test Results

About this item

 :
Boiling Pt:100 °C
Melting Pt:0 °C
Density:0.9971 g/cm3
 :162.14 g/mol
Storage Temperature:Ambient
CAS Number:9005-25-8
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00082026