


Carbon disulfide ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:75-15-0
:
Formula:CS₂
MDL Number:MFCD00011321
:76.14 g/mol
Boiling Pt:46 °C (1.013 hPa)
Melting Pt:–76 °C
Density:1.264 g/cm³ (20 °C)
Flash Pt:–30 °C
Storage Temperature:Ambient
EINECS:200-843-6
UN:1131
ADR:3,I
:GHS02
:GHS08
:GHS07
Specification Test Results
- Identity (IR):complying
- Assay (GC):Min. 99.90 %
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.03 %
- Absorbance at 750 nm:Max. 0.01
- Absorbance 500 nm:Max. 0.01
- Absorbance at 410 nm:Max. 0.05
- Absorbance at 403 nm:Max. 0.10
- Absorbance at 394 nm:Max. 0.25
- Absorbance at 388 nm:Max. 0.50
- Absorbance at 386 nm:Max. 1.0
- Autoignition Temperature:212 °F
- Explosion Limit:50 %
- Vapor Density:2.67 (vs air)