

N,N-Dimethylacetamide ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:127-19-5
:
Formula:CH₃CON(CH₃)₂
MDL Number:MFCD00008686
:87.12 g/mol
Boiling Pt:164…166 °C (1.013 hPa)
Melting Pt:–20 °C
Density:0.94 g/cm³ (20 °C)
Flash Pt:64 °C
Storage Temperature:Ambient
EINECS:204-826-4
UN:2810
ADR:6.1,III
:GHS07
:GHS08
Specification Test Results
- IR spectrum:complying
- Assay (GC):Min. 99.90 %
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.03 %
- Absorbance at 270 nm:Max. 1.0
- Absorbance 280 nm:Max. 0.30
- Absorbance at 290 nm:Max. 0.15
- Absorbance at 310 nm:Max. 0.05
- Absorbance at 320 nm:Max. 0.03
- Absorbance at 360 nm:Max. 0.01
- Absorbance at 400 nm:Max. 0.01
- Explosion Limit:1.8 %, 100 °F
- Vapor Density:3 (vs air)
Specification Test Results
Frequently Bought Together