Order Entry
Puerto Rico
ContactUsLinkComponent
Sodium hypochlorite ∼10% (RT) in aqueous solution, Purum, Fluka™
Corrosive
Sodium hypochlorite ∼10% (RT) in aqueous solution, Purum, Fluka™
Catalog # BJ71696-10L
CAS Number:  
Corrosive
Sodium hypochlorite ∼10% (RT) in aqueous solution, Purum, Fluka™
Catalog # BJ71696-10L
Supplier Number:  71696-10L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    10 L
  • Cat. No.:
    BJ71696-10L
  • Supplier no.:
    71696-10L

Specifications

Specification Test Results

  • Assay of active chlorine:
    8.0 - 15.0 %
  • pH (direct):
    11.5 - 13.5
  • Density (D 20/4):
    1.21 - 1.26

Specification Test Results

About this item

 :
 :74.44 g/mol
Boiling Pt:111 °C
Melting Pt:–30…–20 °C
Density:1.206 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-668-3
UN:1791
ADR:8,II
 :GHS05
CAS Number:7681-52-9
Formula:ClNaO
MDL Number:MFCD00011120