


n-Hexane ≥99.0%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Catalog # BJ52767-1L
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:1 L
- Cat. No.:BJ52767-1L
Specifications
Specification Test Results
- Explosion Limit:7.7 %
- Assay (GC):Min. 99 %
- High-boiling fraction:Max. 1 ppm
- Refractive index (n 20/D):1.374 - 1.376
- Vapor Density:~3 (vs air)
- Vapor Pressure:~132 mmHg ( 20 °C)
- Refractive Index:n20/D 1.375 (lit.)
- Autoignition Temperature:453 °F
Specification Test Results
About this item
:
:86.18 g/mol
Boiling Pt:66…69 °C(1.013 hPa)
Boiling Pt:68.7 °C
Melting Pt:–95 °C
Melting Pt:-95 °C
Density:0.659 - 0.673 g/cm3
Flash Pt:7.4 %(V)
Flash Pt:-26 °C
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
ADR:3,II
:GHS07
:GHS08
:GHS02
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520