

Ethyl acetate ≥99.7% (by GC), CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:141-78-6
:
Formula:CH₃COOC₂H₅
MDL Number:MFCD00009171
:88.11 g/mol
Boiling Pt:77 °C
Melting Pt:-84 °C
Density:0.8945 g/cm3
Flash Pt:-4 °C
Storage Temperature:Ambient
EINECS:205-500-4
UN:1173
ADR:3,II
:GHS07
:GHS02
Specification Test Results
- Assay (GC):Min. 99.7 %
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.03 %
- Free alkali (as NH₃):Max. 0.0005 %
- Calcium (Ca):Max. 0.00001 %
- Potassium (K):Max. 0.00001 %
- Magnesium (Mg):Max. 0.00001 %
- Sodium (Na):Max. 0.00001 %
- Transmittance at 260 nm:Min. 50 %
- Transmittance from 300 nm:Min. 98 %
- Suitability for HPLC-MS:complying
- Vapor Density:3 (20 °C, vs air)
- Autoignition Temperature:801 °F