Order Entry
Puerto Rico
ContactUsLinkComponent
1-Chlorobutane ≥99.8%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Health
Flammable
1-Chlorobutane ≥99.8%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Catalog # BJ34958-2L
CAS Number:  
undefined
Health
Flammable
1-Chlorobutane ≥99.8%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Catalog # BJ34958-2L
Supplier Number:  34958-2L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    2 L
  • Cat. No.:
    BJ34958-2L
  • Supplier no.:
    34958-2L

Specifications

Specification Test Results

  • Assay (GC):
    min. 99.8 %
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Free acid (as HCl):
    Max. 0.001 %
  • Free alkali (as NH₃):
    Max. 0.0002 %
  • Transmittance at 230 nm:
    Min. 65 %
  • Transmittance at 240 nm:
    Min. 95 %
  • Transmittance from 250 nm:
    Min. 98 %

Specification Test Results

About this item

 :
 :92.57 g/mol
Boiling Pt:78.8 °C (1.013 hPa)
Boiling Pt:78 °C
Melting Pt:-123 °C
Density:0.8862 g/cm3
Flash Pt:10.1 %(V)
Flash Pt:-9 °C
Storage Temperature:Ambient
EINECS:203-696-6
UN:1127
ADR:3,II
 :GHS08
 :GHS02
CAS Number:109-69-3
Formula:CH₃(CH₂)₃Cl
MDL Number:MFCD00001009