Acetonitrile ≥99.9% (by GC), CHROMASOLV™ AMD, Riedel-de Haën™
Catalog # BJ34896-1L
Supplier: Honeywell Research Chemicals
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Glass bottle
- Size:1 L
- Environmentally Preferable:
- Cat. No.:BJ34896-1L
- Supplier no.:34896-1L
Specifications
Specification Test Results
- Vapor Density:1.41 (vs air)
- Assay (GC):Min. 99.9 %
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.02 %
- Free acid (as CH₃COOH):Max. 0.001 %
- Free alkali (as NH₃):Max. 0.0001 %
- Transmittance at 195 nm:Min. 75 %
- Transmittance at 200 nm:Min. 93 %
- Transmittance from 230 nm:Min. 99 %
- Baseline drift at 210 nm:Max. 15 mAU
- HPLC Gradient at 210 nm:Max. 3 mAU
- Fluorescence (chinin) at 254 nm:Max. 1 ppb
- Fluorescence (chinin) at 365 nm:Max. 1 ppb
- for AMD technic:complying
- Vapor Pressure:72.8 mmHg ( 20 °C)
- Explosion Limit:16 %
- Refractive Index:n20/D 1.344 (lit.)
Specification Test Results
About this item
Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
:
:41.05 g/mol
Flash Pt:6 °C
EINECS:200-835-2
UN:1648
:GHS08
:GHS07
:GHS02
Storage Temperature:Ambient
CAS Number:75-05-8
Formula:H₃CCN
MDL Number:MFCD00001878
Boiling Pt:81.6
Melting Pt:-45.7
Density:0.7857 g/cm³ (20 °C)
ADR:3 II
Merck Index:13,00071