


Acetonitrile ≥99.8% (by GC), CHROMASOLV™ for liquid chromatography, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:75-05-8
:
Formula:H₃CCN
MDL Number:MFCD00001878
:41.05 g/mol
Boiling Pt:82 °C (1.013 hPa)
Melting Pt:–46 °C
Density:0.7857 g/cm³ (20 °C)
Flash Pt:6 °C
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
:GHS08
:GHS07
:GHS02
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:Chem21 solvent selection guide recommended by default
Specifications
Specification Test Results
- Assay (GC):min. 99.8 %
- Non-volatile matter:max. 0.0005 %
- Water (Karl Fischer):max. 0.1 %
- Free acid (as CH₃COOH):max. 0.001 %
- Transmittance at 200 nm:min. 50 %
- Transmittance at 220 nm:min. 90 %
- Transmittance from 240 nm:min. 99 %