Iron(III) chloride solution 45% (FeCl₃ basis), Purum, Fluka™
BJ12322-2.5L
: Avantor
:
- Pack type:Glass bottle
- Size:2.5 L
- Cat. No.:BJ12322-2.5L
- Supplier no.:12322-2.5L
- Assay (excluding Fe):44.6 - 45.4 %
:
:162.21 g/mol
:1.48 g/cm3
:Ambient
:2582
:8,III
:GHS05
:GHS07
:7705-08-0
:FeCl₃
:MFCD00011005