

Ammonia solution 30 - 33% (NH₃ basis) in water, Puriss., Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MDL Number:MFCD00066650
:35.05 g/mol
Boiling Pt:32 °C (1.013 hPa)
Melting Pt:–92 °C
Density:0.89 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2672
ADR:8,III
:GHS07
:GHS05
Specification Test Results
- Assay:30 - 33 %
- Non-volatile matter:Max. 0.002 %
- Iron (Fe):Max. 0.0001 %
- Heavy metals (as Pb):Max. 0.0003 %
- Chloride (Cl):Max. 0.0002 %
- Sulfate (SO₄):Max. 0.002 %