


Acetonitrile ≥99.9% (by GC), CHROMASOLV™, gradient grade suitable for PAH analysis, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:75-05-8
:
Formula:H₃CCN
MDL Number:MFCD00001878
:41.05 g/mol
Boiling Pt:82 °C
Melting Pt:-46 °C
Density:0.7822 g/cm3
Flash Pt:6 °C
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
:GHS08
:GHS07
:GHS02
Specification Test Results
- Assay (GC):Min. 99.9 %
- Density (D 20/4):0.765 - 0.794
- Refractive index (n 20/D):1.343 - 1.345
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.02 %
- Free acid (as CH₃COOH):Max. 0.005 %
- Free alkali (as NH₃):Max. 0.0005 %
- Suitability test (HPLC):complying
- Absorbance at 230 nm:Max. 0.005
- Absorbance at 215 nm:Max. 0.01
- Absorbance at 200 nm:Max. 0.04
- Fluorescent substances:complying
- Refractive Index:n20/D 1.344 (lit.)