Order Entry
Puerto Rico
ContactUsLinkComponent
 
Corrosive
Sodium hydroxide 5.0 M NaOH (5.0N) in aqueous solution volumetric solution, Fluka™
Catalog # BJ656046-1L
CAS Number:  
Corrosive
Sodium hydroxide 5.0 M NaOH (5.0N) in aqueous solution volumetric solution, Fluka™
Catalog # BJ656046-1L
Supplier Number:  656046-6X1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 L
  • Cat. No.:
    BJ656046-1L

Specifications

Specification Test Results

  • Assay:
    4.900 - 5.100 N
  • Traceable to NIST SRM:
    effective

Specification Test Results

About this item

CAS Number:1310-73-2
 :
Formula:NaOH
MDL Number:MFCD00003548
 :40 g/mol
Boiling Pt:105…140 °C (1.013 hPa)
Melting Pt:–12…–10 °C
Density:1.327 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:1824
ADR:8,II
 :GHS05