
Hydrochloric acid 0.2 M HCl (0.2N) ≥97% volumetric solution, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Boiling Pt:100 °C
Melting Pt:0 °C
Density:1.0 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-791-2
UN:1789
ADR:8,III
:36.46
:GHS05
Specification Test Results
- Appearance (trubidity):complying
- Appearance (color):complying
- Appearance (form):complying
- Traceable to NIST SRM:effective
- Assay:0.1900 - 0.2100 N