
Sodium hydroxide 0.1 M NaOH (0.1N) ≥99% volumetric solution, Fluka™
:1310-73-2
:
:NaOH
:MFCD00003548
:40 g/mol
:100 °C (1.013 hPa)
:319 °C
:1.000 g/cm³ (20 °C)
:Ambient
:231-791-2
:1824
:8,III
:GHS05
- Assay titr.:0.095 - 0.105 mol/l
- Carbonate (CO₃):Max. 0.0015 mol/l
- Traceable to NIST SRM:effective
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective