
Hydrochloric acid 0.1 M HCl (0.1N) 99.5-100.5% volumetric solution, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Boiling Pt:100 °C (1.013 hPa)
Melting Pt:0 °C
Density:1.000 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-791-2
UN:1789
ADR:8,III
:36.46
:GHS05
Specification Test Results
- Assay:0.0950 - 0.1050 N
- Traceable to NIST SRM:effective
- Vapor Pressure:190 mmHg ( 25 °C)