
Hydrochloric acid 1.0 M HCl (1.0N) in aqueous solution volumetric solution, Fluka™
Supplier: Avantor
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Boiling Pt:100 °C
Melting Pt:0 °C
Density:1,010 g/cm3 (20 °C)
Storage Temperature:Ambient
UN:1789
ADR:8,III
:36.46
:GHS05
Specification Test Results
- Factor:0.950 - 1.050
- Traceable to NIST SRM:complying
- Vapor Pressure:190 mmHg ( 25 °C)
Specification Test Results
Related Information
Frequently Bought Together