Order Entry
Puerto Rico
ContactUsLinkComponent
Bromate-Bromide 0.05 M Br2 (0.1 N) volumetric solution, Fluka™
Health
Bromate-Bromide 0.05 M Br2 (0.1 N) volumetric solution, Fluka™
Catalog # BJ35006-1L
CAS Number:  
Health
Bromate-Bromide 0.05 M Br2 (0.1 N) volumetric solution, Fluka™
Catalog # BJ35006-1L
Supplier Number:  35006-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 L
  • Cat. No.:
    BJ35006-1L
  • Supplier no.:
    35006-1L

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002

Specification Test Results

About this item

 :
Density:1.01 g/cm³
 :GHS08
Storage Temperature:Ambient
 :7758-01-2
Formula:KBrO₃/KBr