Order Entry
Puerto Rico
ContactUsLinkComponent
Methyl red solution in ethanol acid-base indicator, Fluka™
Irritant
Flammable
Methyl red solution in ethanol acid-base indicator, Fluka™
Catalog # BJ32941-100ML
CAS Number:  
Irritant
Flammable
Methyl red solution in ethanol acid-base indicator, Fluka™
Catalog # BJ32941-100ML
Supplier Number:  32941-100ML
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass Bottle
  • Size:
    100 ml
  • Cat. No.:
    BJ32941-100ML
  • Supplier no.:
    32941-100ML

Specifications

Specification Test Results

  • Change:
    complying
  • Test of sensitivity:
    complying

Specification Test Results

About this item

 :
Flash Pt:28 °C
EINECS:207-776-1
ADR:3,III
 :269.3 g/mol
UN:1170
 :GHS07
 :GHS02
Storage Temperature:Ambient
CAS Number:493-52-7
Formula:C₁₅H₁₅N₃O₂
MDL Number:MFCD00002425