NeuroMag™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
Certificates
About this item
NeuroMag Transfection Reagent is the first dedicated Magnetofection transfection reagent for neurons and neural cells, from 1 DIV to 21 DIV.
- Great efficiency, ideal for primary neurons
- Efficient from 1 DIV to 21 DIV
- Non toxic and completely biodegradable: high transfected neurons viability
- Ready-to-use, straightforward and rapid
- For all types of nucleic acids
Accessories information: This reagent needs to be used with a magnetic plate.
Delivery information: Shipped at room temperature
Packaging: Store at -20 °C
Specifications
- Cell Specific:Neurons, Glial Cells
- DNA:Yes
- dsRNA:Yes
- miRNA:Yes
- mRNA:Yes
- shRNA:Yes
- siRNA:Yes
Specifications
Frequently Bought Together