PolyMag Neo™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
Certificates
About this item
PolyMag Neo Magnetofection Reagent is specifically designed to achieve high transfection efficiency combined with superior transgene expression level.
- High transgene expression
- High transfection efficiency
- Multipurpose (various types of nucleic acids)
- Universal (primary cells and cell lines)
- Simple, ready-to-use and rapid
- Non toxic
- Compatible with and without serum-containing culture media
It is composed of magnetic nanoparticles coated with cationic molecules.
Accessories information: This reagent needs to be used with a magnetic plate
Delivery information: Shipped at room temperature
Packaging: Store at +4 °C
Specifications
- Cell Specific:Hard-to-transfect
- DNA:Yes
- dsRNA:Yes
- miRNA:Yes
- mRNA:Yes
- shRNA:Yes
- siRNA:Yes
Specifications
Related Information
Frequently Bought Together