Mag4C-AD™ Transfection Reagent, Trial Kit, OZ Biosciences
Mag4C-Ad is designed for magnetic virus concentration.
- Simple, Rapid & Ready-to-use
- Concentration of viruses by magnetic capture in 30-45 minutes
- High yield of viral capture and recovery
- No need to process magnetic beads before capture
- Avoid ultracentrifugation, precipitation and chemicals. Reduced handling steps of viruses
- Suitable for all conditions & viruses
- Improve virus stability under storage conditions
- Ideal for cell culture infection (Magnetofection™ advantages)
This kit is specifically developed for capturing, concentrating and storing adenoviruses.
: OZ Biosciences' multipurpose Magnetic Separation Rack for 50, 15 or 1.5 mL tubes is proposed to be used with the kit.
: Shipped at room temperature
: Store at +4 °C
Frequently Bought Together