si3D-FectIN™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
Certificates
About this item
si3D-FectIN is a 3D transfection reagent specifically designed and developed for silencing gene expression in cells cultured in gels (or hydrogels).
- Highly efficient for gene silencing in 3D matrices
- Ideal for any gel and hydrogel
- Dedicated to short nucleic acid sequences (siRNA, miRNA…)
- Long term gene silencing
- Universal (primary cells and cell lines)
- Serum Compatible
Delivery information: Shipped at room temperature
Packaging: Store at +4 °C
Specifications
- miRNA:Yes
- shRNA:Yes
- siRNA:Yes