DreamFect™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
Certificates
About this item
DreamFect Transfection Reagent is based on an innovative biochemical method.
- Exceptional efficiency
- High reproducibility
- Broad range of cell types
- For all nucleic acids applications
- Non toxic
- Serum compatible
- Simple, rapid, and easy-to-use
It contains both a lipidic part and a cationic part allowing the association of nucleic acids with this transfection reagent and their delivery inside cells. Due to its unique properties, DreamFect releases large DNA amounts in the cytosol increasing DNA nuclear delivery.
Delivery: Shipped at room temperature.
Packaging: Store at +4 °C
Specifications
- Cell Specific:Mammalian, Adherent, Suspension
- DNA:Yes
- dsRNA:Yes
- mRNA:Yes
- shRNA:Yes